Please login to your online account to display your discounted pricing

Alfa Aesar™ Nicotinic hydrazide, 98+%

$55.76 - $195.88

Chemical Identifiers

Chemical Name or Material Nicotinic hydrazide
CAS 553-53-7
Molecular Formula C6H7N3O
Formula Weight 137.14
MDL Number MFCD00006383
Beilstein 119299
Synonym Pyridine-3-carboxylic acid hydrazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0205914

  SDS
alfa aesar™
L0205914
25g Each for $55.76

AAL0205922

  SDS
alfa aesar™
L0205922
100g Each for $195.88
Description & Specifications

Specifications

Formula Weight 137.14
Chemical Name or Material Nicotinic hydrazide
CAS 553-53-7
Assay ≥98%
Molecular Formula C6H7N3O
Beilstein 119299
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number QT1750000
MDL Number MFCD00006383
Quantity 25g
Health Hazard 2 GHS H Statement
H351
Suspected of causing cancer.
Synonym Pyridine-3-carboxylic acid hydrazide
Health Hazard 1 Warning
Melting Point 160° to 163°C

Applications
Nicotinic hydrazide is used in hydrazone library formation. It is used to study the oxidation of isonicotinic acid hydrazide (ionized) by horseradish peroxidase. Nicotinic hydrazide is an inhibitor of peroxidase enzyme. It forms solid metal complexes having strong biological activity

Solubility
Soluble in alcohol, water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351
Suspected of causing cancer.