missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nicotinic Acid Hydrazide, TCI America™

$47.36 - $137.50

Chemical Identifiers

CAS 553-53-7
Molecular Formula C6H7N3O
Molecular Weight (g/mol) 137.142
MDL Number MFCD00006383
InChI Key KFUSANSHCADHNJ-UHFFFAOYSA-N
Synonym nicotinohydrazide, nicotinic acid hydrazide, nicotinic hydrazide, niazid, nicotinylhydrazide, nicotinoyl hydrazine, 3-pyridoyl hydrazine, 3-pyridylcarbonylhydrazine, nicotinoyl hydrazide, hydrazine, nicotinoyl
PubChem CID 11112
IUPAC Name pyridine-3-carbohydrazide
SMILES C1=CC(=CN=C1)C(=O)NN
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N0087100G View Documents TCI America
N0087100G
100g Each for $137.50
 
N008725G View Documents TCI America
N008725G
25g Each for $47.36
 
Specifications

Chemical Identifiers

553-53-7
137.142
KFUSANSHCADHNJ-UHFFFAOYSA-N
11112
C1=CC(=CN=C1)C(=O)NN
C6H7N3O
MFCD00006383
nicotinohydrazide, nicotinic acid hydrazide, nicotinic hydrazide, niazid, nicotinylhydrazide, nicotinoyl hydrazine, 3-pyridoyl hydrazine, 3-pyridylcarbonylhydrazine, nicotinoyl hydrazide, hydrazine, nicotinoyl
pyridine-3-carbohydrazide

Specifications

White
100g
C6H7N3O
nicotinohydrazide, nicotinic acid hydrazide, nicotinic hydrazide, niazid, nicotinylhydrazide, nicotinoyl hydrazine, 3-pyridoyl hydrazine, 3-pyridylcarbonylhydrazine, nicotinoyl hydrazide, hydrazine, nicotinoyl
C1=CC(=CN=C1)C(=O)NN
137.142
137.14
≥98.0% (GC,T)
162°C
553-53-7
MFCD00006383
KFUSANSHCADHNJ-UHFFFAOYSA-N
pyridine-3-carbohydrazide
11112
Crystal-Powder at 20°C
Nicotinic Acid Hydrazide
Safety and Handling

Safety and Handling

RTECSNumber : QT1750000

TSCA : No