missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nicotinic Acid, ≥99%, MP Biomedicals™
Manufacturer: Mp Biomedicals 0210244691
Specifications
Nicotinic Acid | |
White | |
59-67-6 | |
C6H5NO2 | |
3-carboxypyridine, 3-pyridinecarboxylic acid, acidum nicotinicum, akotin, apelagrin, daskil, niacin, nicotinic acid, pellagrin, wampocap | |
C1=CC(=CN=C1)C(=O)O | |
123.111 | |
CHEBI:15940 | |
Solid |
5.7 x 10-6mm Hg at 25°C (estimated)(Lit.) | |
1kg | |
≥99% | |
MFCD00006391 | |
PVNIIMVLHYAWGP-UHFFFAOYSA-N | |
pyridine-3-carboxylic acid | |
938 | |
123.1 |
Chemical Identifiers
59-67-6 | |
123.111 | |
PVNIIMVLHYAWGP-UHFFFAOYSA-N | |
938 | |
pyridine-3-carboxylic acid |
C6H5NO2 | |
MFCD00006391 | |
3-carboxypyridine, 3-pyridinecarboxylic acid, acidum nicotinicum, akotin, apelagrin, daskil, niacin, nicotinic acid, pellagrin, wampocap | |
CHEBI:15940 | |
C1=CC(=CN=C1)C(=O)O |
Safety and Handling
EINECSNumber : 200-441-0
Recommended Storage : Store at room temperature (15° to 30°C)