Please login to your online account to display your discounted pricing

(-)-Nicotine Tartrate (White Solid), Fisher BioReagents


Quantity 1g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP25331

 View more versions of this product

Catalog No. BP25331

  • / Each
  • / Case of 5 Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 462.4
Melting Point 98.00°C
Quantity 1g
TSCA Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.
Physical Form Solid
Molecular Formula C18H26N2O12 · 2 H2O
Color White
Merck Index 15, 6609
Chemical Name or Material (-)-Nicotine Tartrate
Identification Pass Test
Packaging Amber Glass
CAS 65-31-6
ChemAlert Storage Symbol Blue
Specific Rotation Report ° (+ or -)
Synonym [(S)-3-(1-Methyl-2-pyrrolindinyl)pyridine]
Recommended Storage RT

Specifications

Chemical Name or Material (-)-Nicotine Tartrate
CAS 65-31-6
Physical Form Solid
Molecular Formula C18H26N2O12 · 2 H2O
Formula Weight 462.4
Synonym [(S)-3-(1-Methyl-2-pyrrolindinyl)pyridine]
Accessories
 

RT

SDS & Safety Documents