Please login to your online account to display your discounted pricing

(-)-Nicotine Tartrate (White Solid), Fisher BioReagents

$176.88 - $176.88

Chemical Identifiers

Chemical Name or Material (-)-Nicotine Tartrate
CAS 65-31-6
Physical Form Solid
Molecular Formula C18H26N2O12 · 2 H2O
Formula Weight 462.4
Synonym [(S)-3-(1-Methyl-2-pyrrolindinyl)pyridine]
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BP25331

  SDS
fisher scientific company
BP25331
Amber Glass 1g Each for $176.88
Case of 5 Each for $884.40
Description & Specifications

Specifications

Formula Weight 462.4
Melting Point 98.00°C
Quantity 1g
TSCA Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.
Physical Form Solid
Molecular Formula C18H26N2O12 · 2 H2O
Color White
Merck Index 15, 6609
Chemical Name or Material (-)-Nicotine Tartrate
Identification Pass Test
Packaging Amber Glass
CAS 65-31-6
ChemAlert Storage Symbol Blue
Specific Rotation Report ° (+ or -)
Synonym [(S)-3-(1-Methyl-2-pyrrolindinyl)pyridine]
Recommended Storage RT

(-)-Nicotine Tartrate is used in biochemical research applications.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

RT