Please login to your online account to display your discounted pricing

Alfa Aesar™ Nicotinanilide, 98%

$57.57 - $195.88

Chemical Identifiers

Chemical Name or Material Nicotinanilide
CAS 1752-96-1
Molecular Formula C12H10N2O
Formula Weight 198.23
MDL Number MFCD00023572
Beilstein 142066
Synonym Pyridine-3-carboxanilide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0615406

  SDS
alfa aesar™
L0615406
5g Each for $57.57

AAL0615414

  SDS
alfa aesar™
L0615414
25g Each for $195.88
Description & Specifications

Specifications

Formula Weight 198.23
CAS 1752-96-1
Synonym Pyridine-3-carboxanilide
MDL Number MFCD00023572
Assay 98%
Molecular Formula C12H10N2O
Melting Point 119° to 120°C
Beilstein 142066
Quantity 5g
Chemical Name or Material Nicotinanilide