Please login to your online account to display your discounted pricing

Alfa Aesar™ Nicotinamidoxime, 97%

$91.46 - $305.88

Chemical Identifiers

Chemical Name or Material Nicotinamidoxime
CAS 1594-58-7
Molecular Formula C6H7N3O
Formula Weight 137.14
MDL Number MFCD00265955
Synonym 3-Pyridylamidoxime; Pyridine-3-carboxamidoxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5174903

  SDS
alfa aesar™
H5174903
1g Each for $91.46

AAH5174906

  SDS
alfa aesar™
H5174906
5g Each for $305.88
Description & Specifications

Specifications

Formula Weight 137.14
Melting Point 130° to 135°C
Quantity 1g
Health Hazard 1 Warning
MDL Number MFCD00265955
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Nicotinamidoxime
Assay 97%
Molecular Formula C6H7N3O
Synonym 3-Pyridylamidoxime; Pyridine-3-carboxamidoxime
CAS 1594-58-7
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.