missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nicotinamide (Niacinamide), MilliporeSigma™ Supelco™
Description
Environmental and Pharmaceutical (small molecule) Vitamins, Nutraceuticals and Natural Products
Specifications
Specifications
| Packaging | Ampule |
| CAS | 98-92-0 |
| Flash Point | 150°C |
| Physical Form | Neat |
| Linear Formula | C6H6N2 O |
| MDL Number | MFCD00006395 |
| Melting Point | 128°C to 131°C |
| Synonym | Nicotinamide; Niacinamide; Nicotinic acid amide; Pyridine-3-carboxylic acid amide; Vitamin B3; Vitamin PP |
| Type | Organic Pollutant Standard |
| Quantity | 1000 mg |
| Show More |