Please login to your online account to display your discounted pricing

Alfa Aesar™ Nicofuranose

$116.07 - $438.69

Chemical Identifiers

Chemical Name or Material Nicofuranose
CAS 15351-13-0
Molecular Formula C30H24N4O10
Formula Weight 600.52
MDL Number MFCD00868276
Synonym beta-D-Fructofuranose tetranicotinate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2202422

  SDS
alfa aesar™
B2202422
100g Each for $116.07

AAB2202436

  SDS
alfa aesar™
B2202436
500g Each for $438.69
Description & Specifications

Specifications

Formula Weight 600.52
Chemical Name or Material Nicofuranose
Molecular Formula C30H24N4O10
MDL Number MFCD00868276
CAS 15351-13-0
Synonym beta-D-Fructofuranose tetranicotinate
Odor Odorless
Melting Point 133° to 136°C
Quantity 100g