Please login to your online account to display your discounted pricing

Nickelous Sulfate, 6-Hydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$179.13 - $5,186.70

Chemical Identifiers

Chemical Name or Material Nickelous Sulfate
CAS 10101-97-0
Name Note 6-Hydrate
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula NiSO4·6H2O
Formula Weight 262.85
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003455

 
avantor performance
280804
Poly Bottle 125g Each for $179.13

02003457

 
avantor performance
280801
Poly Bottle 500g Each for $476.11

02003456

 
avantor performance
280805
Poly Bottle 2.5kg Each for $5,186.70
Case of 4 Each for $20,746.80
Description & Specifications

Specifications

Chemical Name or Material Nickelous Sulfate
Name Note 6-Hydrate
CAS 10101-97-0
Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Molecular Formula NiSO4·6H2O
Formula Weight 262.85
Packaging Poly Bottle
Specific Gravity 1L = 2.03kg
Quantity 125g