Please login to your online account to display your discounted pricing

Nickelous Chloride, 6-Hydrate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 125g
Packaging Poly Bottle

Manufacturer: avantor performance  276804

 View more versions of this product

Catalog No. 02003453

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystals
Formula Weight 237.71
Chemical Name or Material Nickelous Chloride
CAS 7791-20-0
Name Note 6-Hydrate
Packaging Poly Bottle
Purity Grade BAKER ANALYZED™ Reagent
Specific Gravity 1L = 3.55kg
Molecular Formula NiCl2·6H2O
Quantity 125g

Specifications

Chemical Name or Material Nickelous Chloride
CAS 7791-20-0
Name Note 6-Hydrate
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula NiCl2·6H2O
Formula Weight 237.71