Please login to your online account to display your discounted pricing

Nickelous Chloride, 6-Hydrate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$179.19 - $415.12

Chemical Identifiers

Chemical Name or Material Nickelous Chloride
CAS 7791-20-0
Name Note 6-Hydrate
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula NiCl2·6H2O
Formula Weight 237.71
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003453

 
avantor performance
276804
Poly Bottle 125g Each for $179.19

02003454

 
avantor performance
276801
Poly Bottle 500g Each for $415.12
Description & Specifications

Specifications

Formula Weight 237.71
Physical Form Crystals
Chemical Name or Material Nickelous Chloride
CAS 7791-20-0
Molecular Formula NiCl2·6H2O
Quantity 125g
Name Note 6-Hydrate
Packaging Poly Bottle
Specific Gravity 1L = 3.55kg
Purity Grade BAKER ANALYZED™ Reagent