Please login to your online account to display your discounted pricing

Nickelous Carbonate, Powder, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$745.25 - $1,833.63

Chemical Identifiers

Chemical Name or Material Nickelous Carbonate
CAS 3333-67-3
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula NiCO3
Formula Weight 118.7
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003450

 
avantor performance
276405
Poly Bottle 1.5kg Each for $1,833.63

02003451

 
avantor performance
276401
Poly Bottle 500g Each for $745.25
Description & Specifications

Specifications

Formula Weight 118.7
Quantity 1.5kg
Chemical Name or Material Nickelous Carbonate
Molecular Formula NiCO3
CAS 3333-67-3
Packaging Poly Bottle
Purity Grade BAKER ANALYZED™ Reagent
Physical Form Powder