missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel wire, Annealed, 99.5% (metals basis), Alfa Aesar™

Form: approx. 28.0g/m, Temper: soft

Manufacturer:  Alfa Aesar 14188G5

 View more versions of this product

Catalog No. AA14188G5



Specifications

Specifications

Nickel
7440-02-0
8.908g/cm3 at 20°C
Ni
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
[Ni]
Odorless
935
Wire, 2mm (0.08 in.) dia.
5m
Silver gray
99.5% (Metals basis)
MFCD00011137
PXHVJJICTQNCMI-UHFFFAOYSA-N
nickel
58.693
CHEBI:28112
Silver to Gray

Chemical Identifiers

7440-02-0
58.693
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
nickel
Ni
MFCD00011137
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
CHEBI:28112
[Ni]
Safety and Handling

Safety and Handling

H372-H351-H317: Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction

EINECSNumber : 231-111-4

Documents