missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel wire, 99.994% (metals basis), Alfa Aesar™, Puratronic™

Form: approx. 0.07g/m

Manufacturer:  Thermo Fisher Scientific 10932G2

 View more versions of this product

Catalog No. AA10932G2



Specifications

Specifications

Nickel
7440-02-0
99.994% (Metals basis)
MFCD00011137
PXHVJJICTQNCMI-UHFFFAOYSA-N
nickel
58.693
CHEBI:28112
Silver to Gray
2m
Silver gray wire
Ni
carbonyl powder
[Ni]
Odorless
935
Wire, 0.1mm (0.004 in.) dia.

Chemical Identifiers

7440-02-0
58.693
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
nickel
Ni
MFCD00011137
carbonyl powder
CHEBI:28112
[Ni]
Safety and Handling

Safety and Handling

H372-H351-H317: Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction.

EINECSNumber : 231-111-4

Documents