missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel Wire, 0.5mm (0.02 in.) dia., 99.5% (Metals basis), Alfa Aesar™

Form: Approx.1.75g/m, Temper: soft

Manufacturer:  Alfa Aesar 10250G5

 View more versions of this product

Catalog No. AA10250G5



Specifications

Specifications

Nickel wire
0.5mm (0.02 in.) dia, annealed
≈1.75g/m
Ni
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
[Ni]
Odorless
935
Temper: Soft
5m
7440-02-0
(metals basis)
MFCD00011137
PXHVJJICTQNCMI-UHFFFAOYSA-N
nickel
58.693
CHEBI:28112
99.5%

Chemical Identifiers

7440-02-0
58.693
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
nickel
Ni
MFCD00011137
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
CHEBI:28112
[Ni]
Safety and Handling

Safety and Handling

GHS H Statement
H372-H351-H317;
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.

GHS P Statement
P260-P261-P280-P363-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Documents