Please login to your online account to display your discounted pricing

Nickel wire, 0.5mm (0.02in) dia, annealed, 99.5% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Nickel wire
CAS 7440-02-0
Name Note 0.5mm (0.02 in.) dia, annealed
MDL Number MFCD00011137
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA10250G5

 
alfa aesar
10250G5
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Temper: Soft
Quantity 5m
Chemical Name or Material Nickel wire
TSCA TSCA
Density ≈1.75g/m
Name Note 0.5mm (0.02 in.) dia, annealed
Assay 99.5%
Assay Percent Notes (metals basis)
MDL Number MFCD00011137
CAS 7440-02-0
Odor Odorless
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H372-H351-H317;
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P260-P261-P280-P363-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P260-P261-P280-P363-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
Avoid breathing dust/fume/gas/mist/vapours/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H372-H351-H317;
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.