Learn More
Nickel wire, 0.5mm (0.02in) dia, annealed, 99.5% (metals basis), Thermo Scientific Chemicals
Form: Approx.1.75g/m, Temper: soft
Supplier: Thermo Scientific Chemicals 010250H4
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver to Gray | |
7440-02-0 | |
8.908g/cm3 at 20°C | |
Ni | |
100 m | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
nickel | |
935 | |
Odorless |
Nickel | |
Silver gray wire | |
99.5% (Metals basis) | |
MFCD00011137 MFCD06798735 | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
[Ni] | |
58.69 | |
CHEBI:28112 | |
Wire |
Chemical Identifiers
7440-02-0 | |
58.69 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
935 | |
nickel |
Ni | |
MFCD00011137 MFCD06798735 | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
CHEBI:28112 | |
[Ni] |
Safety and Handling
Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H351-H372
EINECSNumber : 231-111-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only