missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel wire, 0.25mm (0.01 in.) dia., Puratronic™, 99.994% (metals basis)

Manufacturer:  Alfa Aesar™ 10931H2

 View more versions of this product

Catalog No. AA10931H2



Specifications

Specifications

Nickel wire
50m
7440-02-0
Ni
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
[Ni]
Odorless
935
Wire
Puratronic™
≈0.44g/m
0.25mm (0.01 in.) dia.
(metals basis)
MFCD00011137
PXHVJJICTQNCMI-UHFFFAOYSA-N
nickel
58.693
CHEBI:28112
99.994%

Chemical Identifiers

7440-02-0
58.693
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
nickel
Ni
MFCD00011137
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
CHEBI:28112
[Ni]
Safety and Handling

Safety and Handling

GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.

GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Documents