missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nickel wire, 0.25mm (0.01in) dia, annealed, 99.98% (metals basis), Thermo Scientific Chemicals
Form: Approx.0.44g/m, Temper: soft
Supplier: Thermo Scientific Chemicals 43131G5
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver to Gray | |
7440-02-0 | |
Ni | |
5 m | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
nickel | |
935 |
Nickel wire, Annealed | |
99.98% (metals basis) | |
MFCD00011137 MFCD06798735 | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
[Ni] | |
58.69 | |
CHEBI:28112 |
Chemical Identifiers
7440-02-0 | |
58.69 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
935 | |
nickel |
Ni | |
MFCD00011137 MFCD06798735 | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
CHEBI:28112 | |
[Ni] |
Safety and Handling
P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H351-H372
EINECSNumber : 231-111-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only