Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel Vanadium, sputtering target, 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis)

Chemical Identifiers

Chemical Name or Material Nickel Vanadium, sputtering target
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD01310520
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40880KS

 
alfa aesar™
40880KS
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Non-Magnetic
Chemical Name or Material Nickel Vanadium, sputtering target
Name Note 76.2mm (3.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.95%
Assay Percent Notes (metals basis)
MDL Number MFCD01310520
Avg. Mol. Wt. or Mol. Wt. Range Ni:V; 93:7 wt%
Quantity 1 each