Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel tubing, 1.59mm (0.0626in) OD, 0.51mm (0.02in) ID, 99% (metals basis)

Chemical Identifiers

Chemical Name or Material Nickel tubing
CAS 7440-02-0
Name Note OD: 1.59mm (0.0626 in.) ID: 0.51mm (0.02 in.)
MDL Number MFCD00011137
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA13456RS

  SDS
alfa aesar™
13456RS
60cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA13456RT

  SDS
alfa aesar™
13456RT
5 x 60cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Tubes Cut From Coil, May Be Bowed or Bent
Chemical Name or Material Nickel tubing
Assay 99%
Health Hazard 1 Danger
Assay Percent Notes (metals basis)
MDL Number MFCD00011137
Odor Odorless
Health Hazard 3 GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 60cm
Health Hazard 2 GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
Name Note OD: 1.59mm (0.0626 in.) ID: 0.51mm (0.02 in.)
TSCA TSCA
CAS 7440-02-0
Safety and Handling

Danger

GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.