Learn More
Nickel tin oxide dihydrate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03945222
Description
It is used as an active pharmaceutical ingredient.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel tin oxide dihydrate | |
12035-38-0 | |
100 g | |
MFCD00054011 | |
Insoluble in water. | |
225.40 | |
Odorless |
Green | |
Powder | |
NiO3Sn | |
Nickel stannate | |
SHULAURIUQUJKN-UHFFFAOYSA-N | |
261.43 (225.40 Anhydrous) |
Chemical Identifiers
12035-38-0 | |
225.40 | |
SHULAURIUQUJKN-UHFFFAOYSA-N |
NiO3Sn | |
MFCD00054011 | |
Nickel stannate |
Safety and Handling
GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350i-H372
EINECSNumber : 234-824-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only