Please login to your online account to display your discounted pricing

Nickel Thinfoil, 0.006mm (0.0002 in.) thick, hard, 99.95% (Metals basis), Alfa Aesar™

Temper: as rolled; Form: Approx.4.0g/25 x 30cm, not certified pinhole free

Chemical Identifiers

Chemical Name or Material Nickel
Name Note Hard
CAS 7440-02-0
Physical Form Thin Foil, 0.006mm (0.0002 in.) thick
Assay Percent Range 99.95% (Metals basis)
MDL Number MFCD00011137
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA14368FI

 
alfa aesar
14368FI
50 x 50mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA14368GH

 
alfa aesar
14368GH
100 x 100mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA14368HG

 
alfa aesar
14368HG
100 x 300mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Thin Foil, 0.006mm (0.0002 in.) thick
Quantity 50 x 50mm
Color Silver-gray
Chemical Name or Material Nickel
EINECS Number 231-111-4
Name Note Hard
Assay Percent Range 99.95% (Metals basis)
MDL Number MFCD00011137
CAS 7440-02-0
Odor Odorless
Health Hazard 1 H372-H351-H317 : Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction
Appearance Silver gray Thinfoil
Health Hazard 2 Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).
Temper Temper: as rolled
Safety and Handling

H372-H351-H317 : Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. May cause an allergic skin reaction

Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).