missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel sulfide, 99% (metals basis)

Manufacturer:  Alfa Aesar™ 4549814

 View more versions of this product

Catalog No. AA4549814



Specifications

Specifications

Nickel sulfide
(metals basis)
MFCD00016266
nickel iv sulphide
S.S.[Ni]
126.845
Powder
25g
12035-51-7
H4NiS2
UN3077
CBJSXTNBQQMXNY-UHFFFAOYSA-N
nickel;sulfane
131675689
99%

Chemical Identifiers

12035-51-7
126.845
CBJSXTNBQQMXNY-UHFFFAOYSA-N
131675689
S.S.[Ni]
H4NiS2
MFCD00016266
nickel iv sulphide
nickel;sulfane
Safety and Handling

Safety and Handling

GHS H Statement
H350-H372-H341-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.

GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

SDS Documents