missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel Sulfate Hexahydrate (Crystalline/Certified ACS), Fisher Chemical

$250.46 - $527.15

Chemical Identifiers

CAS 10101-97-0
Physical Form Solid
Molecular Formula NiO4S · 6 H2O
Formula Weight 262.85
MDL Number MFCD00149813
Synonym Nickel (II) Sulfate
Chemical Name or Material Nickel Sulfate Hexahydrate
Assay 98 to 102%
Grade Certified ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
N73-100 SDS Fisher Scientific Company
N73100
Poly Bottle 100g Each for $250.46
Case of 6 Each for $1,502.76
N73-500 SDS Fisher Scientific Company
N73500
Poly Bottle 500g Each for $527.15
Case of 6 Each for $3,162.90
Specifications Description & Specifications

Chemical Identifiers

10101-97-0
NiO4S · 6 H2O
MFCD00149813
Nickel Sulfate Hexahydrate
Certified ACS Grade
Solid
262.85
Nickel (II) Sulfate
98 to 102%

Specifications

10101-97-0
NiO4S · 6 H2O
MFCD00149813
0.005% max.
0.002% max.
2.07g/m
0.005% max.
0.005% max.
0.002% max.
0.01% max.
>280°C
Blue Green
4 to 6
Nickel Sulfate Hexahydrate
Certified ACS Grade
Solid
262.85
Nickel (II) Sulfate
0.001% max.
0.005% max.
Pass Test
0.001% max.
0.002% max.
Poly Bottle
0.05% max.
103°C
53°C
100g
98 to 102%

Safety and Handling

WARNING!

Emergency Overview
Cancer hazard. Harmful by inhalation and if swallowed. Irritating to eyes and skin. May cause allergic respiratory and skin reaction. May cause irritation of respiratory tract. May cause harm to the unborn child. Possible risks of irreversible effects. Danger of serious damage to health by prolonged exposure. Very toxic to aquatic organisms