Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.99% (metals basis)

Magnetron sputtering source

$485.18 - $485.18

Chemical Identifiers

Chemical Name or Material Nickel sputtering target
CAS 7440-02-0
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00011137
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40874KS

  SDS
alfa aesar™
40874KS
1 each Each for $485.18
Description & Specifications

Specifications

CAS 7440-02-0
Health Hazard 2 GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Chemical Name or Material Nickel sputtering target
Assay 99.99%
Quantity 1 each
TSCA TSCA
MDL Number MFCD00011137
Assay Percent Notes (metals basis)
Odor Odorless
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.