missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel Sponge, promoted with 1% Molybdenum, , Alfa Aesar™

Manufacturer:  Alfa Aesar 4486922

 View more versions of this product

Catalog No. AA4486922

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Nickel
7697-37-2
HNO3
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
[N+](=O)(O)[O-]
Odorless
944
Sponge, Promoted With 1% Molybdenum
100g
Suspensions
MFCD03844839
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
63.012
CHEBI:48107

Chemical Identifiers

7697-37-2
63.012
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
nitric acid
HNO3
MFCD03844839
hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
CHEBI:48107
[N+](=O)(O)[O-]
Safety and Handling

Safety and Handling

H228-H251-H351-H317 : Flammable solid. Self-heating: may catch fire. Suspected of causing cancer. May cause an allergic skin reaction

DOTInformation : DOT Class: 4.2, Packaging Group: II

SDS Documents