Please login to your online account to display your discounted pricing

Nickel Sponge, promoted with 1% Molybdenum, , Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Nickel
Physical Form Sponge, Promoted With 1% Molybdenum
MDL Number MFCD03844839
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4486922

 
alfa aesar
4486922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4486936

 
alfa aesar
4486936
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Sponge, Promoted With 1% Molybdenum
Quantity 100g
DOT Information DOT Class: 4.2, Packaging Group: II
Chemical Name or Material Nickel
MDL Number MFCD03844839
Odor Odorless
Health Hazard 1 H228-H251-H351-H317 : Flammable solid. Self-heating: may catch fire. Suspected of causing cancer. May cause an allergic skin reaction
Appearance Suspensions
Safety and Handling

H228-H251-H351-H317 : Flammable solid. Self-heating: may catch fire. Suspected of causing cancer. May cause an allergic skin reaction