missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel on silica-alumina, catalyst, Alfa Aesar™

Formula: 66±5% Ni

Manufacturer:  Alfa Aesar 3127606

 View more versions of this product

Catalog No. AA3127606



Specifications

Specifications

Nickel
on silica-alumina, catalyst
Black catalyst powder
MFCD00011137
PXHVJJICTQNCMI-UHFFFAOYSA-N
[Ni]
58.693
CHEBI:28112
Black
5g
7440-02-0
Ni
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
Odorless
nickel
935
Catalyst Powder

Chemical Identifiers

7440-02-0
58.693
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
nickel
Ni
MFCD00011137
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
CHEBI:28112
[Ni]
Safety and Handling

Safety and Handling

H228-H351-H332-H317-H335: Flammable solid. Suspected of causing cancer. Harmful if inhaled. May cause an allergic skin reaction. May cause respiratory irritation.

DOTInformation : DOT Class: 4.1, Packaging Group: II

SDS Documents