missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel Shots, 3-25mm (0.1-0.98 in.), >99.95% (Metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1025130

 View more versions of this product

Catalog No. AA1025130



Specifications

Specifications

Nickel
7440-02-0
8.908g/cm3 at 20°C
Ni
carbonyl powder, catalyst, fibrex, fibrex p, niccolum, nichel italian, nickel, elemental, nickel, soluble salts, particles, raney alloy
[Ni]
Odorless
935
Shots, 3-25mm (0.1-0.98 in.)
250g
Silver gray shot
>99.95% (Metals basis)
MFCD00011137
PXHVJJICTQNCMI-UHFFFAOYSA-N
nickel
58.693
CHEBI:28112
Silver to Gray

Chemical Identifiers

7440-02-0
58.693
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
nickel
Ni
MFCD00011137
carbonyl powder, catalyst, fibrex, fibrex p, niccolum, nichel italian, nickel, elemental, nickel, soluble salts, particles, raney alloy
CHEBI:28112
[Ni]
Safety and Handling

Safety and Handling

Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H351-H372

EINECSNumber : 231-111-4

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents