Please login to your online account to display your discounted pricing

Nickel, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical


Quantity 100mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  SN70100

 View more versions of this product

Catalog No. SN70-100

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Boiling Point 100°C
Quantity 100mL
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Color Blue Green
Vapor Pressure 14mmHg at 20°C
Viscosity [NULL]
Concentration 990 to 1010ppm
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Chemical Name or Material Nickel, Reference Standard Solution
Name Note 1000ppm ±1%
MDL Number MFCD00011137
CAS 13478-00-7
ChemAlert Storage Symbol White
CAS 7697-37-2
Packaging Poly Bottle
Traceability to NIST LOT NUMBER USED NICKEL REF. STANDARD
CAS 7732-18-5

Specifications

Chemical Name or Material Nickel, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 13478-00-7
CAS 7697-37-2
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00011137
Accessories
 

SDS & Safety Documents