Please login to your online account to display your discounted pricing

Nickel, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

$76.71 - $108.12

Chemical Identifiers

Chemical Name or Material Nickel, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 13478-00-7
CAS 7697-37-2
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00011137
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SN70-100

  SDS
fisher scientific company
SN70100
Poly Bottle 100mL Each for $76.71
Case of 6 Each for $460.26

SN70-500

  SDS
fisher scientific company
SN70500
Poly Bottle 500mL Each for $108.12
Case of 6 Each for $648.72
Description & Specifications

Specifications

Chemical Name or Material Nickel, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 13478-00-7
ChemAlert Storage Symbol White
CAS 7697-37-2
CAS 7732-18-5
DOT Information DOT Class 8, : Corrosive
Purity Grade Certified Grade
Physical Form Liquid
Concentration 990 to 1010ppm
MDL Number MFCD00011137
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Packaging Poly Bottle
Traceability to NIST LOT NUMBER USED NICKEL REF. STANDARD
Vapor Pressure 14mmHg at 20°C
Boiling Point 100°C
Color Blue Green
Quantity 100mL
Safety and Handling