missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel Powder, spherical, APS 0.08 to 0.15 micron, 99.8% (Metals basis), Alfa Aesar™

Form: S.A. 4.5-8.5m2/g, Tap density 1.0-2.5g/cm3

Manufacturer:  Alfa Aesar 4333830

 View more versions of this product

Catalog No. AA4333830



Specifications

Specifications

Nickel
7440-02-0
99.8% (Metals basis)
MFCD00011137
PXHVJJICTQNCMI-UHFFFAOYSA-N
nickel
58.693
CHEBI:28112
Silver to Gray
250g
Silver gray powder
Ni
carbonyl powder
[Ni]
Odorless
935
Spherical Powder

Chemical Identifiers

7440-02-0
58.693
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
nickel
Ni
MFCD00011137
carbonyl powder
CHEBI:28112
[Ni]
Safety and Handling

Safety and Handling

H372-H351-H228-H317 : Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. Flammable solid. May cause an allergic skin reaction

DOTInformation : DOT Class: 4.1, Packaging Group: II

EINECSNumber : 231-111-4

Documents