Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel powder, spherical, -325 mesh (5% max. +325), 99.7% min. (metals basis)b

Chemical Identifiers

Chemical Name or Material Nickel
CAS 7440-02-0
MDL Number MFCD00011137
UN Number UN3089
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA77725P2

 
alfa aesar™
77725P2
1 lb. CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Spherical Powder
Chemical Name or Material Nickel
TSCA TSCA
Assay ≥99.7%
Assay Percent Notes (metals basis)
MDL Number MFCD00011137
CAS 7440-02-0
UN Number UN3089
Mesh Size −325 mesh (5% max. +325)
Odor Odorless
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H228-H250-H351-H317
Flammable solid.
Catches fire spontaneously if exposed to air.
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P210-P222-P363-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not allow contact with air.
Wash contaminated clothing before reuse.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.1; Packaging Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
RTECS Number QR5950000
Quantity 1 lb.
Safety and Handling

Danger

GHS P Statement
P210-P222-P363-P405-P422a-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not allow contact with air.
Wash contaminated clothing before reuse.
Store locked up.
Store contents under inert gas.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H250-H351-H317
Flammable solid.
Catches fire spontaneously if exposed to air.
Suspected of causing cancer.
May cause an allergic skin reaction.