missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel Powder, Spherical, -300 mesh, 99.8% (Metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1058136

 View more versions of this product

Catalog No. AA1058136



Specifications

Specifications

Nickel
7440-02-0
8.908g/cm3 at 20°C
Ni
carbonyl powder
[Ni]
Odorless
935
Spherical Powder
500g
Silver gray powder of flakes
99.8% (Metals basis)
MFCD00011137
PXHVJJICTQNCMI-UHFFFAOYSA-N
nickel
58.693
CHEBI:28112
Silver to Gray

Chemical Identifiers

7440-02-0
58.693
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
nickel
Ni
MFCD00011137
carbonyl powder
CHEBI:28112
[Ni]
Safety and Handling

Safety and Handling

Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets.

H372-H351-H228-H317 : Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. Flammable solid. May cause an allergic skin reaction

DOTInformation : DOT Class: 4.2, Packaging Group: II

EINECSNumber : 231-111-4

Documents