missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel powder, low carbon, 99.999% (metals basis), Alfa Aesar™, Puratronic™

C less-than100ppm

Manufacturer:  Alfa Aesar 1296614

 View more versions of this product

Catalog No. AA1296614



Specifications

Specifications

Nickel
7440-02-0
99.999% (Metals basis)
MFCD00011137
PXHVJJICTQNCMI-UHFFFAOYSA-N
nickel
58.693
CHEBI:28112
Silver to Gray
25g
Silver gray powder of flakes
Ni
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
[Ni]
Odorless
935
Low Carbon Powder

Chemical Identifiers

7440-02-0
58.693
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
nickel
Ni
MFCD00011137
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
CHEBI:28112
[Ni]
Safety and Handling

Safety and Handling

H372-H351-H228-H317: Causes damage to organs through prolonged or repeated exposure. Suspected of causing cancer. Flammable solid. May cause an allergic skin reaction

DOTInformation : DOT Class: 4.1, Packaging Group: II

EINECSNumber : 231-111-4

Documents