missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nickel Powder, 75% on graphite, <75 micron, Alfa Aesar™

Form: Apparent density 0.8-1.5g/cm3

Manufacturer:  Thermo Fisher Scientific 1055536

 View more versions of this product

Catalog No. AA1055536

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Nickel
7440-02-0,7782-42-5
HNO3
acide nitrique
[N+](=O)(O)[O-]
Odorless
944
Powder, 75% On Graphite
500g
Black powder
MFCD00011137
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
63.012
CHEBI:48107
Black

Chemical Identifiers

7440-02-0
63.012
GRYLNZFGIOXLOG-UHFFFAOYSA-N
944
nitric acid
HNO3
MFCD00011137
acide nitrique
CHEBI:48107
[N+](=O)(O)[O-]
Safety and Handling

Safety and Handling

H228-H251-H351-H319-H317-H335 : Flammable solid. Self-heating: may catch fire. Suspected of causing cancer. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation

DOTInformation : DOT Class: 4.2, Packaging Group: II

Documents