Please login to your online account to display your discounted pricing

Nickel Powder, 75% on graphite, <75 micron, Alfa Aesar™

Form: Apparent density 0.8-1.5g/cm3

Chemical Identifiers

Chemical Name or Material Nickel
Physical Form Powder, 75% On Graphite
MDL Number MFCD00011137
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1055522

 
alfa aesar
1055522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1055536

 
alfa aesar
1055536
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder, 75% On Graphite
Quantity 100g
Color Black
DOT Information DOT Class: 4.2, Packaging Group: II
Chemical Name or Material Nickel
MDL Number MFCD00011137
Odor Odorless
Health Hazard 1 H228-H251-H351-H319-H317-H335 : Flammable solid. Self-heating: may catch fire. Suspected of causing cancer. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation
Appearance Black powder
Safety and Handling

H228-H251-H351-H319-H317-H335 : Flammable solid. Self-heating: may catch fire. Suspected of causing cancer. Causes serious eye irritation. May cause an allergic skin reaction. May cause respiratory irritation