Learn More
Nickel powder, -325 mesh, typically 99.8% (metals basis), Thermo Scientific Chemicals
Form: Conductive dendritic powder
Supplier: Thermo Scientific Chemicals 043214A3
Description
Conductive surface coatings, inks, adhesives, and sealants
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver to Gray | |
7440-02-0 | |
99.8% (Metals basis) | |
MFCD00011137 MFCD06798735 | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
[Ni] | |
58.69 | |
CHEBI:28112 | |
Odorless |
Nickel | |
Silver gray powder | |
Ni | |
2 kg | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
nickel | |
935 | |
-325 Mesh | |
Conductive Dendritic Powder |
Chemical Identifiers
7440-02-0 | |
58.69 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
935 | |
nickel |
Ni | |
MFCD00011137 MFCD06798735 | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
CHEBI:28112 | |
[Ni] |
Safety and Handling
Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets.
P201-P202-P210-P240-P241-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c
H228-H317-H350-H372
DOTInformation : DOT Class: 4.1, Packaging Group: II
EINECSNumber : 231-111-4
RTECSNumber : QR5950000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only