Learn More
Nickel, plasma standard solution, Specpure™ Ni 1000μg/mL, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 013839AP
Description
Nickel, plasma standard solution is used as a standard solution in analytical chemistry and atomic absorption spectroscopy. It is also used as a single-element standard solution for plasma emission spectrometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel | |
500 mL | |
Ni | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
nickel | |
935 | |
Nitric acid: 5.0%; Nickel: 0.1%; Water: 94.9% | |
Miscible with water |
7440-02-0,7697-37-2,7732-18-5 | |
Colorless | |
MFCD00011137 MFCD06798735 | |
[Ni] | |
58.69 | |
CHEBI:28112 | |
23 hPa (17mm Hg) at 20°C | |
Liquid |
Chemical Identifiers
7440-02-0 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
CHEBI:28112 | |
[Ni] |
58.69 | |
935 | |
nickel |
Safety and Handling
P201-P202-P234-P260-P264b-P271-P281-P285-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H290-H314-H335-H350i-H360D
DOTInformation : Hazard Class: 8; Packaging Group: II
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only