Please login to your online account to display your discounted pricing

Nickel Oxide, Green (Powder/Certified), Fisher Chemical


Quantity 500g
Packaging Glass Bottle

Manufacturer: fisher scientific company  N69500

 View more versions of this product

Catalog No. N69-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Physical Form Powder/Solid
Formula Weight 74.69
Melting Point 1960°C
Quantity 500g
Viscosity [NULL]
Color Black
Density 6.670g/cm³
Sodium (Na) 0.02% max.
Sulfate 0.01% max.
Iron (Fe) 0.01% max.
Lead (Pb) 0.005% max.
Nitrogen Compounds 0.005% max.
Packaging Glass Bottle
Potassium (K) 0.005% max.
MDL Number MFCD00011145
CAS 1313-99-1
ChemAlert Storage Symbol Blue
Synonym Nickel (II) Oxide
Calcium (Ca) 0.005% max.
Chloride 0.005% max.
Chemical Name or Material Nickel Oxide, Green
Identification Pass Test
Zinc (Zn) 0.02% max.
Cobalt (Co) 0.2% max.
Copper (Cu) 0.02% max.
Molecular Formula NiO

Specifications

Chemical Name or Material Nickel Oxide, Green
CAS 1313-99-1
Purity Grade Certified Grade
Physical Form Powder/Solid
Molecular Formula NiO
Formula Weight 74.69
MDL Number MFCD00011145
Synonym Nickel (II) Oxide
Accessories
 

SDS & Safety Documents