Please login to your online account to display your discounted pricing

Nickel Oxide, Green (Powder/Certified), Fisher Chemical

$549.00 - $1,195.46

Chemical Identifiers

Chemical Name or Material Nickel Oxide, Green
CAS 1313-99-1
Purity Grade Certified Grade
Physical Form Powder/Solid
Molecular Formula NiO
Formula Weight 74.69
MDL Number MFCD00011145
Synonym Nickel (II) Oxide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

N69-100

  SDS
fisher scientific company
N69100
Glass Bottle 100g Each for $549.00
Case of 6 Each for $3,294.00

N69-500

  SDS
fisher scientific company
N69500
Glass Bottle 500g Each for $1,195.46
Case of 6 Each for $7,172.76
Description & Specifications

Specifications

Purity Grade Certified Grade
Physical Form Powder/Solid
Formula Weight 74.69
Melting Point 1960°C
Quantity 100g
Viscosity [NULL]
Color Black
Density 6.670g/cm³
Chemical Name or Material Nickel Oxide, Green
Identification Pass Test
Potassium (K) 0.005% max.
Sulfate 0.01% max.
CAS 1313-99-1
ChemAlert Storage Symbol Blue
Synonym Nickel (II) Oxide
Calcium (Ca) 0.005% max.
Copper (Cu) 0.02% max.
Iron (Fe) 0.01% max.
Nitrogen Compounds 0.005% max.
Packaging Glass Bottle
Sodium (Na) 0.02% max.
MDL Number MFCD00011145
Zinc (Zn) 0.02% max.
Chloride 0.005% max.
Cobalt (Co) 0.2% max.
Lead (Pb) 0.005% max.
Molecular Formula NiO
Safety and Handling