Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel naphthenate, Ni 5-12%

Chemical Identifiers

Chemical Name or Material Nickel naphthenate
CAS 61788-71-4
MDL Number MFCD00147723
UN Number UN3082
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA39455A1

  SDS
alfa aesar™
39455A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Viscous Liquid
Chemical Name or Material Nickel naphthenate
TSCA TSCA
MDL Number MFCD00147723
CAS 61788-71-4
UN Number UN3082
Color Green
Quantity 1kg
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P201-P308+P313
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Concentration Ni 5 to 12%
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P201-P308+P313
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.