missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel nanopowder, APS 5-20nm, 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 4550514

 View more versions of this product

Catalog No. AA4550514



Specifications

Specifications

Nickel nanopowder
7440-02-0
Ni
UN3089
PXHVJJICTQNCMI-UHFFFAOYSA-N
[Ni]
58.693
CHEBI:28112
Powder
Black
25g
(metals basis)
MFCD00011137
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
Odorless
nickel
935
APS 5 to 20nm, NiO layer 0.5 to 1.5nm
99.9%

Chemical Identifiers

7440-02-0
58.693
PXHVJJICTQNCMI-UHFFFAOYSA-N
935
nickel
Ni
MFCD00011137
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
CHEBI:28112
[Ni]
Safety and Handling

Safety and Handling

GHS H Statement
H372-H351-H228-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Flammable solid.
May cause an allergic skin reaction.

GHS P Statement
P210-P260-P261-P363-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

RTECSNumber : QR5950000

SDS Documents