missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel molybdenum oxide, 98%

Manufacturer:  Alfa Aesar™ 8993822

 View more versions of this product

Catalog No. AA8993822



Specifications

Specifications

Nickel molybdenum oxide
MoNiO4
molybdenum nickel tetraoxide, nickel molybdate, nickel 2+ molybdate, molybdenum nickel oxide, nickel 2+ dioxido dioxo molybdenum, moo4.ni, nickel molybdenum oxide, dioxido dioxo molybdenum; nickel 2+
[O-][Mo](=O)(=O)[O-].[Ni+2]
218.639
218.65
98%
100g
14177-55-0
MFCD00016255
NLPVCCRZRNXTLT-UHFFFAOYSA-N
dioxido(dioxo)molybdenum;nickel(2+)
84241
Powder
Odorless

Chemical Identifiers

14177-55-0
218.639
NLPVCCRZRNXTLT-UHFFFAOYSA-N
84241
[O-][Mo](=O)(=O)[O-].[Ni+2]
MoNiO4
MFCD00016255
molybdenum nickel tetraoxide, nickel molybdate, nickel 2+ molybdate, molybdenum nickel oxide, nickel 2+ dioxido dioxo molybdenum, moo4.ni, nickel molybdenum oxide, dioxido dioxo molybdenum; nickel 2+
dioxido(dioxo)molybdenum;nickel(2+)
Safety and Handling

Safety and Handling

GHS H Statement
H350-H372-H317
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
May cause an allergic skin reaction.

GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

SDS Documents