missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Alfa Aesar™ Nickel lump, vacuum remelted, low Oxygen, 99.97% (metals basis)
Manufacturer: Alfa Aesar™ 1295722
Specifications
Nickel lump, vacuum remelted | |
Low Oxygen, Oxygen and Nitrogen analysis included | |
(metals basis) | |
MFCD00011137 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
nickel | |
58.693 | |
CHEBI:28112 |
100g | |
7440-02-0 | |
Ni | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
[Ni] | |
Odorless | |
935 | |
99.97% |
Chemical Identifiers
7440-02-0 | |
58.693 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
935 | |
nickel |
Ni | |
MFCD00011137 | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
CHEBI:28112 | |
[Ni] |
Safety and Handling
GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
GHS P Statement
P260-P261-P280-P363-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
TSCA : TSCA