Please login to your online account to display your discounted pricing

Alfa Aesar™ Nickel Iron, sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.95% (metals basis)

Magnetron sputtering source

Chemical Identifiers

Chemical Name or Material Nickel Iron, sputtering target
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD01741928
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40886KS

  SDS
alfa aesar™
40886KS
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Nickel Iron, sputtering target
TSCA TSCA
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.95%
Assay Percent Notes (metals basis)
MDL Number MFCD01741928
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Avg. Mol. Wt. or Mol. Wt. Range Ni:Fe; 81:19 wt%
Quantity 1 each
Safety and Handling

Warning

GHS P Statement
P261-P280-P281-P363-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
Wash contaminated clothing before reuse.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.