Alfa Aesar™ Nickel Iron Molybdenum flake, -200 mesh
Manufacturer:  Alfa Aesar™ 4561030
 View more versions of this product
Catalog No. AA4561030
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications
Specifications
Ni:Fe:Mo; 81:17:2 wt%
250g
MFCD09910537
−200 Mesh
Nickel Iron Molybdenum flake
0.29g/mL
UN3089
Flakes
Documents