Learn More
Nickel(II) tetrafluoroborate hexahydrate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 05310836
Description
It is mainly used as an electroplating intermediate. It is also used in medical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel (II) tetrafluoroborate hexahydrate | |
Lump | |
B2F8H12NiO6 | |
UN3077 | |
Soluble in water. | |
O.O.O.O.O.O.[Ni++].F[B-](F)(F)F.F[B-](F)(F)F | |
340.39 | |
340.40 (232.30 Anhydrous) | |
Hygroscopic |
15684-36-3 | |
500 g | |
MFCD00150259 | |
nickel ii tetrafluoroborate hexahydrate, nickel ii ditetrafluoroborate hexahydrate, nickel 2+ ditetrafluoroborate hexahydrate, borate 1-,tetrafluoro-, nickel 2+ 2:1 , hexahydrate 9ci, 2bf4.ni.6h2o, nickel ii tetrafluoroboratehexahydrate, nickel ii tetrafluoroborate hexahydrate 100g, nickel 2+ hexahydrate ditetrafluoroborate, nickel 2+ ion hexahydrate ditetrafluoroborate | |
BFFWGFMAXHCLDV-UHFFFAOYSA-N | |
nickel(2+);ditetrafluoroborate;hexahydrate | |
177615 | |
Odorless | |
1.5 g/mL |
Chemical Identifiers
15684-36-3 | |
340.39 | |
BFFWGFMAXHCLDV-UHFFFAOYSA-N | |
177615 |
B2F8H12NiO6 | |
MFCD00150259 | |
nickel ii tetrafluoroborate hexahydrate, nickel ii ditetrafluoroborate hexahydrate, nickel 2+ ditetrafluoroborate hexahydrate, borate 1-,tetrafluoro-, nickel 2+ 2:1 , hexahydrate 9ci, 2bf4.ni.6h2o, nickel ii tetrafluoroboratehexahydrate, nickel ii tetrafluoroborate hexahydrate 100g, nickel 2+ hexahydrate ditetrafluoroborate, nickel 2+ ion hexahydrate ditetrafluoroborate | |
O.O.O.O.O.O.[Ni++].F[B-](F)(F)F.F[B-](F)(F)F |
Safety and Handling
GHS H Statement
H334-H350-H360-H372-H341-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H350
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 238-753-4
RTECSNumber : QR6650000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only